Learn More
Phospho-MEK1 (Ser218/222)/MEK2 (Ser222/226) Rabbit anti-Human, Mouse, Rat, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | phospho-MEK1 (Ser218/222)/MEK2 (Ser222/226) |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Formulation | 0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30% |
| Gene Accession No. | NM_002755, NM_030662 |
| Host Species | Rabbit |
| Immunogen | Peptide containing DpSMANpSF in which pS corresponds to phosphorylated amino acids 218 and 222 of human MEK1 |
| Purification Method | Protein A purfied |
| Quantity | 100 μL |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.