Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-phospho Mis18-binding protein 1 (Thr702), Polyclonal

Catalog No. ABE1847MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ABE1847MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE1847MI Supplier MilliporeSigma™ Supplier No. ABE1847
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects phospho Mis18-binding protein 1 (Thr702) Clone: in Human samples, and it is validated for Dot Blot, Immunocytochemistry, Western Blotting

Mis18-binding protein 1 (UniProt Q6P0N0; also known as hsKNL-2, Kinetochore null 2 homolog, Kinetochore-associated protein KNL-2 homolog, P243, Putative protein p243 which interacts with transcription factor Sp1) is encoded by the MIS18BP1 (also known as C14orf106, KIAA1903, KNL2, M18BP1) gene (Gene ID 55320) in human. Mis18-binding protein 1 is a component of the Mis18 complex (Mis18α, Mis18β, and M18BP1/KNL2) that plays a role in mediating cell cycle-dependent deposition/incorporation of the histone H3 variant CENP-A at the centromeres. CENP-A incorporation is carried out by at least two sets of events, the Mis18 complex must first localize to centromeres at the onset of anaphase, HJURP-mediated CENP-A deposition then occurs during G1. Polo-like kinase 1 (Plk1) is shown to phosphorylate M18BP1 on Thr702 and plays an important role in promoting Mis18 complex centromere localization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho Mis18-binding protein 1 (Thr702)
Applications Dot Blot, Immunocytochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in PBS with 0.05% sodium azide and 30% glycerol.
Gene Accession No. Q6P0N0
Gene Symbols MIS18BP1, C14orf106, KIAA1903, KNL2, M18BP1
Host Species Rabbit
Immunogen Linear peptide corresponding to a human M18BP1 sequence containing phosphorylated Thr702.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Epigenetics, Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_060823
Test Specificity Specifically reacts with the immunogen peptide, but not the corresponding non-phosphorylated sequence, by dot blot analysis. Phosphorylation site is not present in the spliced isoform 2 of human Mis18-binding protein 1/M18BP1.
Target Species Human
Content And Storage −20°C for one year
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.