Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Phospho-MKK7/SKK4 (Thr275) Rabbit anti-Human, Polyclonal

Catalog No. 36013MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-013-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 36-013-MI Supplier MilliporeSigma™ Supplier No. 36013

Rabbit Polyclonal Antibody

This Anti-phospho-MKK7/SKK4 (Thr275) Antibody is validated for use in western blotting for the detection of phospho-MKK7/SKK4 (Thr275).
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-MKK7/SKK4 (Thr275)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation 0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30%
Gene Symbols MAP2K7; MKK7; PRKMK7; MEK7; MAPKK7; JNKK2; Jnkk2
Host Species Rabbit
Immunogen peptide corresponding to amino acids surrounding phosphorylated threonine 275 of human MKK7/SKK4
Purification Method Protein A purfied
Quantity 200 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_145185.2
Target Species Human
Content And Storage −20°C for 24 months
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.