Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-MST1/MST2 (Thr183, Thr180) Rabbit anti-Drosophila, Guinea Pig, Human, Mouse, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA517674
Description
It is not recommended to aliquot this antibody. This antibody is not cross-reactive with other unrelated proteins.

Specifications
Phospho-MST1/MST2 (Thr183, Thr180) | |
Polyclonal | |
STK4 | |
Mst1, Mst2, Serine/threonine-protein kinase 4, Mammalian STE20-like protein kinase 1, MST-1, STE20-like kinase MST1, Serine/threonine-protein kinase Krs-2, Serine/threonine-protein kinase 4 37kDa subunit, MST1/N, Serine/threonine-protein kinase 4 18kDa subunit, MST1/C, STK4, KRS2, MST1, Serine/threonine-protein kinase 3, Mammalian STE20-like protein kinase 2, MST-2, STE20-like kinase MST2, Serine/threonine-protein kinase Krs-1, Serine/threonine-protein kinase 3 36kDa subunit, MST2/N, Serine/threonine-protein kinase 3 20kDa subunit, MST2/C, STK3, KRS1, MST2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100715685, 100729700, 311622, 56274, 58231, 65189, 6788, 6789 | |
-20°C | |
Liquid |
Western Blot | |
Unconjugated | |
Q13043, Q13188, Q9JI10, Q9JI11, O54748 | |
STK3, STK4 | |
Synthetic phosphopeptide corresponding to residues surrounding pThr183 of human Mst1 | |
100 μL | |
Primary | |
Drosophila, Guinea Pig, Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction