Learn More
Phospho-mTOR (Ser2448) Rabbit anti-Human, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | phospho-mTOR (Ser2448) |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Purified rabbit polyclonal IgG in storage buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. |
| Gene Symbols | FRAP1; RAPT1; mTOR; FRAP; MTOR; FLJ44809; FRAP2; RAFT1; FRAP1 FK506 binding protein 12-rapamycin associated protein 1; mammalian target of rapamycin |
| Host Species | Rabbit |
| Immunogen | Amino acids encompassing and including phosphorylated Ser2448 of human mTOR |
| Purification Method | Affinity Purified |
| Quantity | 100 μg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.