Learn More
Phospho-mTOR (Thr2446) Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma 09345
Description
Anti-mTOR Antibody detects level of phospho-mTOR (Thr2446) and has been published and validated for use in western blotting.
Specifications
phospho-mTOR (Thr2446) | |
Polyclonal | |
Affinity purified rabbit IgG in buffer containing 0.1M Tris-Glycine, 0.15M NaCl, 0.05% Sodium Azide, pH7.4. | |
Rabbit | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat |
Western Blot | |
Unconjugated | |
mammalian target of rapamycin; FRAP1 FK506 binding protein 12-rapamycin associated protein 1; RAFT1; FRAP2; FLJ44809; MTOR; FRAP; mTOR; RAPT1; FRAP1 | |
Amino acids encompassing and including phosphorylated Ser2446 of human mTOR | |
100 μL | |
NP_004949 | |
2-8°C for up to 12 months |