Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-Munc-18 (Ser515) Rabbit anti-Rat, Polyclonal, MilliporeSigma™

Catalog No. AB9842MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB9842MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB9842MI Supplier MilliporeSigma Supplier No. AB9842
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This Anti-phospho-Munc-18 (Ser515) Antibody is validated for use in western blotting for the detection of phospho-Munc-18 (Ser515).
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-Munc-18 (Ser515)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Liquid in 10mM HEPES (pH 7.5), 150mM NaCl, 100 ug/mL BSA and 50% glycerol.
Gene Symbols Stxbp1; Unc18a
Host Species Rabbit
Immunogen Synthetic peptide corresponds to sequence surrounding the phosphoserine 515 site of rat Munc-18.
Purification Method Affinity Purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_013038.3
Target Species Rat
Content And Storage −20°C in undiluted aliquotes for 6 months. Avoid repeated freeze/thaw cycles.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.