Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-MYPT1 (Thr850) Rabbit anti-Chicken, Human, Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. 36003MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-003-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 36-003-MI Supplier MilliporeSigma Supplier No. 36003
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect phospho-MYPT1 (Thr850) using this Anti-phospho-MYPT1 (Thr850) Antibody validated for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-MYPT1 (Thr850)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Protein A Purified rabbit IgG in a solution of 30% glycerol, 0.07M Tris-glycine, pH 7.4, 0.135M NaCl, and 0.035% sodium azide as a preservative.
Gene Symbols PPP1R12A; MBS; MGC133042; MYPT1
Host Species Rabbit
Immunogen Peptide corresponding to amino acid residues 845-855 of chicken MYPT1; this sequence is identical to amino acids 848-858 of human MYPT1, with the phosphorylated Thr at position 853.
Purification Method Protein A purfied
Quantity 200 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_002480.1
Target Species Chicken, Human, Mouse, Rat
Content And Storage −20°C in undiluted aliquotes for 24 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.