Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-Neurogranin (Ser36)/Neuromodulin (Ser41) Rabbit anti-Mouse, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma ABN426
Description
This Anti-phospho-Neurogranin (Ser36)/Neuromodulin (Ser41) Antibody is validated for use in Western Blotting for the detection of phospho-Neurogranin (Ser36)/Neuromodulin (Ser41).
Specifications
phospho-Neurogranin (Ser36)/Neuromodulin (Ser41) | |
Polyclonal | |
Rabbit Polyclonal serum with 0.05% sodium azide. | |
Rabbit | |
Unpurified | |
Primary | |
Mouse | |
Serum |
Western Blot | |
Unconjugated | |
Nrgn; neurogranin; Ng; Neurogranin; hng; RC3 | |
KLH-conjugated linear peptide corresponding to the IQ domain of Rat phospho-Neurogranin. | |
100 μg | |
NP_077054 | |
−20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction