Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-p38 MAPK alpha (Thr180, Tyr182), Polyclonal, Invitrogen™

Catalog No. PIPA126679
Change view
Click to view available options
Quantity:
50 μL
Catalog No. Quantity
PIPA126679 50 μL
1 options

Catalog No. PIPA126679

Supplier: Thermo Scientific PA126679

Rabbit Polyclonal Antibody

PA1-26679 detects p38 (phospho T180 + Y182) from human and rat samples. PA1-26679 is expected to cross react with mouse, canine, monkey, and carp (100% conserved).PA1-26679 has been successfully used in Western blot procedures.The PA1-26679 immunogen is a synthetic phosphopeptide corresponding to a portion of human p38 containing threonine 180 and tyrosine 182. This portion is conserved among many species including human, rat, mouse, dog, and carp.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-p38 MAPK pThr180+Tyr182
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Gene MAPK14
Gene Accession No. Q16539, P70618
Gene Symbols MAPK14
Host Species Rabbit
Immunogen Synthetic phosphopeptide corresponding to a portion of human p38 containing threonine 180 and tyrosine 182. This portion is conserved among many speci
Purification Method Antigen affinity chromatography
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1432
Target Species Human, Rat
Content And Storage Store at -80°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.