Learn More
Phospho-p38 MAPK (Thr180/Tyr182) Mouse anti-Human, AlexaFluor™488, Clone: 28B10, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | phospho-p38 MAPK (Thr180/Tyr182) |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 28B10 |
| Conjugate | Alexa Fluor 488 |
| Host Species | Mouse |
| Quantity | 100 Tests |
| Primary or Secondary | Primary |
| Target Species | Human |
| Content And Storage | 2-8°C in undiluted aliquots for up to 6 months. Protect from light. |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.