Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-p38α (Thr180/Tyr182) Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. 09272MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-272-MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-272-MI Supplier MilliporeSigma Supplier No. 09272
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This Anti-phospho-p38α (Thr180/Tyr182) Antibody is validated for use in immunoprecipitation, western blotting for the detection of phospho-p38α (Thr180/Tyr182).
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-p38α (Thr180/Tyr182)
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal serum in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl, and 0.05% sodium azide.
Gene Symbols MAPK14; PRKM14; RK; SAPK2A; CSBP1; p38; CSPB1; CSBP; PRKM15; Mxi2; p38ALPHA; MXI2; EXIP; CSBP2
Host Species Rabbit
Immunogen Peptide corresponding to amino acid region encompassing the human, mouse, and rat phospho-p38α (Thr180/Tyr182).
Purification Method Affinity Purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_001315.1; NM_139012.1; NM_139013.1; NM_139014.1
Target Species Human, Mouse, Rat
Content And Storage −20°C in undiluted aliquotes for 24 months. Avoid repeated freeze/thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.