Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-P70S6K (Thr470) Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. ABS431MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABS431MI 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABS431MI Supplier MilliporeSigma Supplier No. ABS431

Rabbit Polyclonal Antibody

Anti-phospho-P70S6K (Thr470) Antibody is a highly specific rabbit polyclonal antibody, that targets S6 Kinase and has been tested in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-P70S6K (Thr470)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Symbols RPS6KB1; STK14A
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to the auto inhibitory domain of human P70S6K phosphorylated at Thr470.
Purification Method Affinity Purified
Quantity 200 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_003152
Target Species Human
Content And Storage 2-8°C for 12 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.