Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-PACSIN1/Syndapin 1 (Ser346) Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. ABS39MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABS39MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABS39MI Supplier MilliporeSigma Supplier No. ABS39

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Polyclonal Antibody

Anti-phospho-PACSIN1/Syndapin 1 (Ser346) Antibody is an antibody against phospho-PACSIN1/Syndapin 1 (Ser346) for use in Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-PACSIN1/Syndapin 1 (Ser346)
Applications Western Blot
Classification Polyclonal
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Purified rabbit polyclonal serum in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Symbols PACSIN1
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to human PACSIN1/Syndapin 1 phosphorylated at Ser346.
Purification Method Affinity Purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_065855
Target Species Human
Content And Storage 2-8°C for 12 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.