Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-PDGFRα (Tyr754) Rabbit anti-Mouse, Human, Rat, Bovine, Polyclonal, MilliporeSigma™

Catalog No. ABS150MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABS150MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABS150MI Supplier MilliporeSigma Supplier No. ABS150

Rabbit Polyclonal Antibody

Detect phospho-PDGFRα (Tyr754) using this Anti-phospho-PDGFRα (Tyr754) Antibody validated for use in Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-PDGFRα (Tyr754)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Symbols PDGFRA; PDGFR2; RHEPDGFRA
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to human PDGFRα phosphorylated at (Tyr754).
Purification Method Affinity Purified
Quantity 200 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_006197
Target Species Bovine, Human, Mouse, Rat
Content And Storage 2-8°C for 12 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.