Learn More
MilliporeSigma™ Phospho-Phospho-lamban (Ser16) Rabbit anti-Mouse, Rat, Human, Bovine, Canine, Pig, Rabbit, Polyclonal
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | phospho-Phospholamban (Ser16) |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Purified rabbit polyclonal IgG in buffer containing 0.014M phosphate buffer, pH 7.6, 0.175M NaCl, 0.07% sodium azide and 30% glycerol. Store at −20°C. |
| Gene Symbols | CMD1P; OTTHUMP00000017092; PLB; phospholamban |
| Host Species | Rabbit |
| Immunogen | KLH-conjugated, synthetic peptide corresponding to amino acids 14-25 of human cardiac phospholamban (RA[pS]TIEMPQQAR-C) with an added C-terminal cysteine. The immunizing sequence is identical in mouse, rat, rabbit, pig and dog. |
| Purification Method | Affinity Purified |
| Quantity | 100 μg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.