Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-phospho-Phospholamban (Ser16); Polyclonal
Supplier: MilliporeSigma™ 07052/DEL
Description
Mayer, E.J., et al., BBRC, 267: 40, 2000
Western Blotting
Specifications
| phospho-Phospholamban (Ser16) | |
| Unconjugated | |
| KLH-conjugated, synthetic peptide corresponding to amino acids 14-25 of human cardiac phospholamban (RA[pS]TIEMPQQAR-C) | |
| 100 μg | |
| Cell Signaling, Endocrine/Metabolism | |
| Bovine, Canine, Human, Mouse, Pig, Rabbit, Rat |
| Polyclonal | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction