Learn More
Phospho-PKCß II (Thr641) Rabbit anti-Human, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | phospho-PKCβ II (Thr641) |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Purified rabbit Polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. |
| Gene Symbols | PRKCB1; PRKCB; PRKCB2; MGC41878; PKCB; PKC-B; PKC-beta |
| Host Species | Rabbit |
| Immunogen | KLH-conjugated linear peptide corresponding to the AGC-kinase C-terminal domain of Human PKC Beta II phosphorylated at Thr641. |
| Purification Method | Affinity Purified |
| Quantity | 100 μL |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.