Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-PKCδ (Ser664) Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. 07875MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-875-MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-875-MI Supplier MilliporeSigma Supplier No. 07875
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect phospho-PKCδ (Ser664) using this Anti-phospho-PKCδ (Ser664) Antibody validated for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-PKCδ (Ser664)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation 100μL of affinity purified rabbit polyclonal IgG in 50% storage buffer (PBS (without Mg2+ and Ca2+), pH 7.3 containing 1.0mg/mL BSA (IgG, protease free) and 0.05% sodium azide) and 50% glycerol.
Gene Symbols PRKCD; MGC49908; nPKC-delta; MAY1
Host Species Rabbit
Immunogen Peptide corresponding to amino acid region encompassing the human phospho-PKCδ (Ser664).
Purification Method Affinity Purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_006254.3; NM_212539.1
Target Species Human
Content And Storage -20°C in undiluted aliquotes for up to 24 months, Avoid Freeze/Thaw Cycles
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.