Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-PLCγ-1 Rabbit anti-Human, Mouse, Canine, Horse, Rat, Polyclonal, MilliporeSigma™

Catalog No. 072134MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-213-4MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-213-4MI Supplier MilliporeSigma Supplier No. 072134

Rabbit Polyclonal Antibody

This phospho-PLCγ-1 antibody is validated for use in western blotting for the detection of the phospho-PLCγ-1 protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-PLCγ-1 (Tyr783)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal serum in buffer containing 0.1M Tris-glycine, pH 7.4, 150mM NaCl and 0.05% sodium azide.
Gene Symbols 1-phosphatidyl-D-myo-inositol-4,5-bisphosphate ; EC 3.1.4.11 ; OTTHUMP00000031787; OTTHUMP00000178982; PLC-148 ; PLC-II ; PLC-gamma-1; PLC1 ; PLC148 ; PLCgamma1; inositoltrisphosphohydrolase
Host Species Rabbit
Immunogen KLH conjugated linear peptide corresponding to phospho-PLCγ-1 (Tyr783).
Purification Method Affinity Purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_002651
Target Species Canine, Equine, Human, Mouse, Rat
Content And Storage 2-8°C for 6 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.