Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-PRAS40 (Thr246) (Proline-Rich AKT substrate) Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™

Catalog No. 07888MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-888-MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-888-MI Supplier MilliporeSigma Supplier No. 07888
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-phospho-PRAS40 (Thr246) (Proline-Rich AKT substrate) Antibody is an antibody against phospho-PRAS40 (Thr246) for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-PRAS40 (Thr246) (Proline-Rich AKT substrate)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal IgG in buffer containing PBS (without Mg2+ and Ca2+)), pH 7.3 containing 1.0mg/mL BSA (IgG, protease free) and 0.05% sodium azide and 50% glycerol.
Gene Symbols Lobe; MGC2865; OTTHUMP00000196757; OTTHUMP00000196758; OTTHUMP00000196759; OTTHUMP00000196760; PRAS40
Host Species Rabbit
Immunogen Peptide corresponding to amino acid region encompassing the human phospho-PRAS40 (Thr246).
Purification Method Affinity Purfied
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_001092102
Target Species Human, Mouse
Content And Storage -20°C in undiluted aliquots for up to 12 months, Avoid Freeze/Thaw Cycles
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.