Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-phospho PTEN (Ser385), Polyclonal

Catalog No. 07890IMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
07-890-IMI 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-890-IMI Supplier MilliporeSigma™ Supplier No. 07890I
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects phospho PTEN (Ser385) Clone: in Human samples, and it is validated for Peptide Inhibition Assay, Western Blotting

Phosphatidylinositol 3,4,5-trisphosphate 3-phosphatase and dual-specificity protein phosphatase PTEN (EC 3.1.3.16, EC 3.1.3.48, EC 3.1.3.67; UniProt P60484; also known as Mitochondrial PTENalpha, Phosphatase and tensin-like protein, Mutated in multiple advanced cancers 1, Mitochondrial phosphatase and tensin protein alpha, MMAC1 phosphatase and tensin homolog deleted on chromosome 10, Phosphatidylinositol-3,4,5-trisphosphate 3-phosphatase and dual-specificity protein phosphatase PTEN) is encoded by the PTEN (also known as 10q23del, BZS, CWS1, DEC, GLM2, TEP1, MMAC1, MHAM, PTEN1) gene (Gene ID 5728) in human. PTEN protein catalyzes the removal of phosphate from phosphoserine, phosphothreonine, and phosphotyrosine residues on substrate proteins, as well as the phosphate in the D3 position of the inositol ring from phosphatidylinositol 3,4,5-trisphosphate, phosphatidylinositol 3,4-diphosphate, phosphatidylinositol 3-phosphate and inositol 1,3,4,5-tetrakisphosphate. PTEN plays a major role in controlling cell growth and proliferation, and PTEN gene mutains are found in a large number of cancers at high frequency. PTEN activity, stability, localization, and conformation are controlled through complex regulatory mechanisms, including the reversible phosphorylations of several Ser/Thr (S/T) residues in its unfolded C-terminal tail region (PTEN-Ctail, residues 351–403). PTEN S370, S380, T382, T383 and S385 are known target sites by casein kinase II (CK2), while S361, T363, S362, and T366 are phosphorylated by glycogen synthase kinase-3β (GSK3β). In addition, Polo-like kinase 1 (Plk1), Plk3, the microtubule associated serine/threonine (MAST) kinase family, the serine/threonine kinase 11 (STK11, LKB1), and the casein kinase 1 (CK1) have also been reported to phosphorylate PTEN.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho PTEN (Ser385)
Applications Inhibition Assays, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. P60484
Gene Symbols PTEN, MMAC1, TEP1
Host Species Rabbit
Immunogen Peptide corresponding to amino acid region encompassing the human, mouse, and rat phospho-PTEN (Ser385).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_000305
Test Specificity PTEN phosphorylated on Serine residue 385
Target Species Human
Content And Storage 2°C to 8°C for one year
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.