Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-Raf-1 (Ser338) Mouse anti-Mouse, MilliporeSigma™

Catalog No. 05538MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-538-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-538-MI Supplier MilliporeSigma Supplier No. 05538
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-phospho-Raf-1 (Ser338) Antibody detects level of phospho-Raf-1 (Ser338) and has been published and validated for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-Raf-1 (Ser338)
Applications Western Blot
Classification Monoclonal
Conjugate Unconjugated
Formulation Protein A Purified immunoglobulin in Protein A Purified immunoglobulin in 0.1M Tris-Glycine, 0.15M NaCl, 0.05% sodium azide, pH 7.4.
Gene Accession No. NM_002880
Gene Symbols RAF1, cRaf, Raf-1, c-Raf, RAF, CRAF, C-RAF
Host Species Mouse
Immunogen Peptide corresponding to residues 333-345 of human Raf-1
Purification Method Protein G purfied
Quantity 200 μg
Primary or Secondary Primary
Target Species Mouse
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.