Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-Raptor (Ser877) Rabbit anti-Human, Primate, Monkey, Polyclonal, MilliporeSigma™

Catalog No. 09107MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-107-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-107-MI Supplier MilliporeSigma Supplier No. 09107

Rabbit Polyclonal Antibody

This Anti-phospho-Raptor (Ser877) Antibody is validated for use in western blotting for the detection of phospho-Raptor (Ser877).
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-Raptor (Ser877)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit serum in PBS Other with 50% Glycerol.
Gene Symbols raptor; Raptor
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to human Raptor phosphorylated at and around Ser877.
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_065812
Target Species Human, Monkey, Primate
Content And Storage −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.