Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-RNA Pol II (Ser2) Rat anti-Human, Monkey, Canine, Mouse, Rat, Clone: 3E7C7, MilliporeSigma™

Catalog No. MABE953MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE953MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE953MI Supplier MilliporeSigma Supplier No. MABE953
Add to Cart
Add to Cart

Rat Monoclonal Antibody

This Anti-phospho RNA Pol II (Ser2) antibody, clone 3E7C7 is validated for use in western blotting, ChIP-seq, ELISA, immunocytochemistry and Chimmunoprecipitation for the detection of phospho RNA Pol II (Ser2).
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho RNA Pol II (Ser2)
Applications ChIP Assay, ELISA, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 3E7C7
Conjugate Unconjugated
Formulation Purified rat monoclonal IgG2aκ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_000928
Gene Symbols POLR2A, POLR2
Host Species Rat
Immunogen KLH-conjugated linear peptide corresponding to human RNA Pol II phosphorylated at Ser2.
Purification Method Protein G purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Canine, Human, Monkey, Mouse, Rat
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.