Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-RNA Pol II (Ser7) Rat anti-Human, Mouse, Canine, Monkey, Rat, Clone: 3D4A12, MilliporeSigma™

Catalog No. MABE955MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE955MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE955MI Supplier MilliporeSigma Supplier No. MABE955
Add to Cart
Add to Cart

Rat Monoclonal Antibody

This Anti-phospho RNA Pol II (Ser7) antibody, clone 3D4A12 is validated for use in western blotting, ChIP-seq, Chimmunoprecipitation, immunocytochemistry and ELISA for the detection of phospho RNA Pol II (Ser7).
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho RNA Pol II (Ser7)
Applications ChIP Assay, ELISA, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 3D4A12
Concentration 1mg/mL
Conjugate Unconjugated
Formulation Purified rat monoclonal IgG2bκ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_000928
Gene Symbols POLR2A
Host Species Rat
Immunogen KLH-conjugated linear peptide corresponding to human phospho RNA Pol II (Ser7).
Purification Method Protein G purified
Quantity 200 μg
Primary or Secondary Primary
Target Species Canine, Human, Monkey, Mouse, Rat
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.