Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-RNAPII (Tyr1) Rat anti-Human, Clone: 3D12, MilliporeSigma™

Catalog No. MABE350MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE350MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE350MI Supplier MilliporeSigma Supplier No. MABE350
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Detect the phospho-RNAPII (Tyr1) protein using this Anti-phospho-RNAPII (Tyr1), clone 3D12 validated for use in western blot, immunoprecipitation and Chimmunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-RNAPII (Tyr1)
Applications ChIP Assay, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 3D12
Conjugate Unconjugated
Formulation Purified rat monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_000928
Gene Symbols POLR2A, POLR2
Host Species Rat
Immunogen Ovalbumin-conjugated linear peptide corresponding to human RNAPII phosphorylated at Tyr1.
Purification Method Protein G purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.