Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-S6 (Ser235, Ser236), Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Supplier:  Thermo Scientific PA125544

Catalog No. PIPA125544


Description

Description

Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a cytoplasmic ribosomal protein that is a component of the 40S subunit. The protein belongs to the S6E family of ribosomal proteins. It is the major substrate of protein kinases in the ribosome, with subsets of five C-terminal serine residues phosphorylated by different protein kinases. Phosphorylation is induced by a wide range of stimuli, including growth factors, tumor-promoting agents, and mitogens. Dephosphorylation occurs at growth arrest. The protein may contribute to the control of cell growth and proliferation through the selective translation of particular classes of mRNA. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.

PA1-25544 specifically detects human RPS6 phosphorylated at serines 235 and 236, and does not cross react with RPS6 phosphorylated at serines 244 and 247. PA1-25544 is expected to cross react with mouse and rat due to sequence homology.PA1-25544 has been successfully used in Western blot procedures.The PA1-25544 immunogen is a synthetic phosphopeptide corresponding to a portion of human RPS6 that contains serines 235 and 236.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Phospho-RPS6 pSer235+236
Polyclonal
RPS6
RP11-513M16.6
Rabbit
Antigen affinity chromatography
RUO
6194
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Purified
Western Blot
Unconjugated
P62753, P62754, P62755
RPS6
Synthetic phosphopeptide corresponding to a portion of human RPS6 that contains serines 235 and 236.
50 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.