Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-RSK3 (S218) Rabbit anti-Human, Mouse, Polyclonal, R&D Systems™
Rabbit Polyclonal Antibody
Supplier: R&D Systems AF893
This item is not returnable.
View return policy
Description
Phospho-RSK3 (S218) Polyclonal antibody AF893 Specifically detects Phospho-RSK3 (S218) in Human, Mouse samples. It is validated for Immunohistochemistry, Western BlottingSpecifications
RSK3 (p ser218) | |
Polyclonal | |
Western Blot 0.5 ug/mL, Immunohistochemistry 5-15 ug/mL | |
RPS6KA2 | |
Phosphopeptide containing human and mouse RSK3 S218 site | |
100 μg | |
Primary | |
Detects human and mouse RSK3 when phosphorylated at S218. Reactivity with other RSK isoforms when phosphorylated at corresponding sites is unknown. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC, as supplied. 1 month, 2 to 8 degreesC under sterile conditions after opening. 6 months, -20 to -70 degreesC under sterile conditions after opening. |
Western Blot, Immunohistochemistry | |
Unconjugated | |
EC 2.7.11, EC 2.7.11.1, HU-2, MAP kinase-activated protein kinase 1c, MAPK-activated protein kinase 1c, MAPKAP kinase 1c, MAPKAPK-1c, MAPKAPK1C, p90-RSK 2, p90RSK2, p90-RSK3, ribosomal protein S6 kinase alpha 2, ribosomal protein S6 kinase alpha-2, ribosomal protein S6 kinase, 90kD, polypeptide 2, ribosomal protein S6 kinase, 90kDa, polypeptide 2, Ribosomal S6 kinase 3,90 kDa ribosomal protein S6 kinase 2, RPS6KA2, RSK, RSK-3, RSK3pp90RSK3, S6K-alpha, S6K-alpha2, S6K-alpha-2 | |
Rabbit | |
Affinity Purified | |
RUO | |
6196 | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction