Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-Ser/Thr-Pro MPM-2 Mouse anti-Vertebrates, Clone: MPM-2, MilliporeSigma™

Catalog No. 05368MGMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-368-MGMI 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-368-MGMI Supplier MilliporeSigma Supplier No. 05368MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect phospho-Ser/Thr-Pro MPM-2 also known as Mitotic protein #2 with Anti-phospho-Ser/Thr-Pro MPM-2 Antibody (Mouse Monoclonal Antibody), that has been demonstrated to work in ELISA, immunohistochemistry, immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-Ser/Thr-Pro MPM-2
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone MPM-2
Conjugate Unconjugated
Formulation Protein G Purified immunoglobulin in Protein G Purified immunoglobulin in mouse IgG in 1.0 mL PBS containing no preservatives.
Host Species Mouse
Immunogen Mitotic human HeLa cell cytosolic lysate
Purification Method Protein G purfied
Quantity 1 mg
Primary or Secondary Primary
Target Species Vertebrates
Content And Storage −20°C in undiluted aliquots for up to 24 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.