Learn More
Phospho-Ser/Thr-Pro MPM-2 Mouse anti-Vertebrates, Clone: MPM-2, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | phospho-Ser/Thr-Pro MPM-2 |
| Applications | ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | MPM-2 |
| Conjugate | Unconjugated |
| Formulation | Protein G Purified immunoglobulin in Protein G Purified immunoglobulin in 30% glycerol, 0.07M Tris-glycine, pH 7.4, 0.105MNaCl, 0.035% sodium azide as a preservative. |
| Host Species | Mouse |
| Immunogen | Mitotic human HeLa cell cytosolic lysate |
| Purification Method | Protein G purfied |
| Quantity | 200 μg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.