Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-Ser/Thr-Pro MPM-2 Mouse anti-Vertebrates, Cy5, MilliporeSigma™

Catalog No. 16220MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-220-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-220-MI Supplier MilliporeSigma Supplier No. 16220
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-phospho-Ser/Thr-Pro Antibody, MPM-2, Cy5 conjugate is a Mouse Monoclonal Antibody for detection of phospho-Ser/Thr-Pro also known as Mitotic protein #2 and has been tested in immunofluoresence, western blot, immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-Ser/Thr-Pro MPM-2
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Conjugate Cyanine5
Formulation 100μg Cy5-conjugated mouse IgG1 in 200μL of PBS containing 1% BSA, 0.05% Tween™-20, 0.05% sodium azide.
Host Species Mouse
Immunogen Mitotic human HeLa cell lysate
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Vertebrates
Content And Storage 4°C for one year
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.