Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-Ser/Thr-Pro MPM-2 Mouse anti-All, FITC, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 16155
Description
Detect phospho-Ser/Thr-Pro also known as Mitotic protein #2 with Anti-phospho-Ser/Thr-Pro Antibody, MPM-2, FITC conjugate (Mouse Monoclonal Antibody), that has been demonstrated to work in immunocytochemistry.
Specifications
phospho-Ser/Thr-Pro MPM-2 | |
Monoclonal | |
TBS, pH 8.0, 0.05% sodium azide and 30% glycerol | |
200 μg | |
All species |
Immunocytochemistry | |
FITC | |
Mouse | |
Primary | |
−20°C for up to 12 months. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction