Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-SGK Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. 36002MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-002-MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 36-002-MI Supplier MilliporeSigma Supplier No. 36002
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-phospho-SGK Antibody is an antibody against phospho-SGK for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-SGK
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation UnPurified whole rabbit antiSerum containing 30% glycerol and 0.035% sodium azide.
Gene Symbols SGK; SGK1
Host Species Rabbit
Immunogen peptide corresponding to amino acids surrounding Ser255/Thr256 of human SGK1.
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_005627.2
Target Species Human
Content And Storage −20°C in undiluted aliquotes for 24 months. Avoid repeated freeze/thaw cycles.
Form Serum
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.