Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-Smad1 (Ser463/465) Rabbit anti-Human, Mouse, Rat, Xenopus, Polyclonal, MilliporeSigma™

Catalog No. 06702MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-702-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-702-MI Supplier MilliporeSigma Supplier No. 06702
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect phospho-Smad1 (Ser463/465) with Anti-phospho-Smad1 (Ser463/465) Antibody (Rabbit Polyclonal Antibody), that has been shown to work in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-Smad1 (Ser463/465)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Protein A Purified immunoglobulin in 0.1M Tris-glycine, pH 7.4, 0.15M NaCl with 0.05% sodium azide as a preservative.
Gene Symbols SMAD1; MADR1; JV41; JV4-1; GARS; CMT2D; hSMAD1; HMN5; MADH1; BSP-1; BSP1
Host Species Rabbit
Immunogen peptide corresponding to (CGGSPHNPISpSVpSGG) corresponding to amino acids 455-465 of human Smad1 protein
Purification Method Protein A purfied
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_005900
Target Species Human, Mouse, Rat, Xenopus species
Content And Storage −20°C in undiluted aliquotes for 24 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.