Learn More
Phospho-Smad2 (Ser465/467) Rabbit anti-Canine, Human, Mouse, Zebrafish, Clone: A5S, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | phospho-Smad2 (Ser465/467) |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | A5S |
| Conjugate | Unconjugated |
| Formulation | Cultured supernantant in 0.05% sodium azide |
| Gene Accession No. | NM_005901.4, NM_001003652.2 |
| Host Species | Rabbit |
| Immunogen | KLH-conjugated, synthetic peptide corresponding to amino acids [pS]M[pS] surrounding sites Ser465/467 of human Smad2 |
| Quantity | 100 μL |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.