Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-STAT1 (Ser727) Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™

Catalog No. 07714MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-714-MI 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-714-MI Supplier MilliporeSigma Supplier No. 07714

Rabbit Polyclonal Antibody

Detect phospho-STAT1 (Ser727) also known as signal transducer and activator of transcription 1 with Anti-phospho-STAT1 (Ser727) Antibody (rabbit polyclonal antibody), that has been demonstrated to work in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-STAT1 (Ser727)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Gene Symbols STAT1; ISGF-3; STAT91; DKFZp686B04100
Host Species Rabbit
Immunogen KLH-conjugated, synthetic peptide containing the sequence (PM[pS]PE), in which [pS] corresponds to phospho-serine at residue 727 of human STAT1. The immunizing sequence is identical in mouse, rat, canine and chimpanzee.
Purification Method Serum
Quantity 200 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_007315.2; NM_139266.1
Target Species Human, Mouse
Content And Storage −20°C for 24 months
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.