Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-STAT2 (Tyr689) Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™

Catalog No. ABE541MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABE541MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE541MI Supplier MilliporeSigma Supplier No. ABE541
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-phospho-STAT2 (Tyr689) Antibody is an antibody against phospho-STAT2 (Tyr689) for use in Immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-STAT2 (Tyr689)
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Symbols STAT2
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to mouse STAT2 phosphorylated at Tyr689.
Purification Method Affinity Purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_064347
Target Species Human, Mouse
Content And Storage 2-8°C for 12 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.