Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-STAT3 (Tyr705) Mouse anti-Human, Mouse, Canine, Clone: 9E12, MilliporeSigma™

Catalog No. 05485MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-485-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-485-MI Supplier MilliporeSigma Supplier No. 05485
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Use Anti-phospho-STAT3 (Tyr705) Antibody, clone 9E12 (Mouse Monoclonal Antibody) validated in ELISA, immunoprecipitation, western blotting to detect phospho-STAT3 (Tyr705) also known as Acute-phase response factor or DNA-binding protein APRF. This Stat3 Antibody has been validated in Human, Mouse and Canine samples.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-STAT3 (Tyr705)
Applications ELISA, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 9E12
Conjugate Unconjugated
Formulation Lyophilized purified mouse IgG1ќ from 1 mL 2x PBS / 0.09 % Na-azide / PEG and Sucrose. Reconstitute with 100μL H2O (15 min, RT) for a final concentration of 1mg/mL.
Gene Accession No. NM_139276
Gene Symbols STAT3, APRF, FLJ20882, MGC16063
Host Species Mouse
Immunogen KLH-conjugated phosphopeptide containing the AP[pY]LK motif of STAT3
Purification Method Thiophilic adsoption and size exclusion chromatography
Quantity 100 μg
Primary or Secondary Primary
Target Species Canine, Human, Mouse
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.