Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-STAT5A/B (Tyr694/699) Mouse anti-Human, Mouse, Rat, Bovine, Sheep, Clone: 8-5-2, MilliporeSigma™

Catalog No. 05495MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-495-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-495-MI Supplier MilliporeSigma Supplier No. 05495
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-phospho-STAT5A/B (Tyr694/699) Antibody, clone 8-5-2 is a mouse monoclonal antibody for detection of phospho-STAT5A/B (Tyr694/699) also known as signal transducer and activator of transcription 5A/B and has been validated in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-STAT5A/B (Tyr694/699)
Applications Western Blot
Classification Monoclonal
Clone 8/5/2002
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG in buffer containing PBS. Frozen solution at −20°C.
Gene Accession No. NM_003152.2
Gene Symbols STAT5A, MGF, STAT5
Host Species Mouse
Immunogen KLH-conjugated, synthetic phosphopeptide (KAVDG[pY]VKPQIK) corresponding to amino acids 689-700 of STAT5A and 694-705 of STAT5B. Clone 8-5-2.
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Bovine, Human, Mouse, Rat, Sheep
Content And Storage Stable for 1 year at from date of receipt.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.