Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-STAT6 (Tyr641) Mouse anti-Human, Mouse, Clone: 19E2.1, MilliporeSigma™

Catalog No. MABE270MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE270MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE270MI Supplier MilliporeSigma Supplier No. MABE270
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-phospho STAT6 (Tyr641) Antibody, clone 19E2.1 is a mouse monoclonal antibody for detection of phospho STAT6 (Tyr641) also known as Signal transducer and activator of transcription 6, IL-4 Stat and has been validated in western blot, flow cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho STAT6 (Tyr641)
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone 19E2.1
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2aκ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_001171549
Gene Symbols STAT6
Host Species Mouse
Immunogen KLH-conjugated linear peptide corresponding to human STAT6 phosphorylated at (Tyr641).
Purification Method Protein G purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.