Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-Syk (Tyr323), Polyclonal, Invitrogen™

Catalog No. PIPA125543
Change view
Click to view available options
Quantity:
50 μL
Catalog No. Quantity
PIPA125543 50 μL
1 options

Catalog No. PIPA125543

Supplier: Thermo Scientific PA125543

Rabbit Polyclonal Antibody

PA1-25543 detects Syk (phospho Y323) from human samples. PA1-25543 is expected to cross react with mouse (79% conserved) due to sequence homology.PA1-25543 has been successfully used in Western blot procedures.The PA1-25543 immunogen is a synthetic phosphopeptide corresponding to a region of human Syk, containing tyrosine 323 (tyrosine 317 in the mouse sequence).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-Syk pTyr323
Applications Western Blot
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Gene SYK
Gene Accession No. P43405, P48025
Gene Alias DKFZp313N1010, FLJ25043, FLJ37489
Gene Symbols SYK
Host Species Rabbit
Immunogen Synthetic phosphopeptide corresponding to a region of human Syk, containing tyrosine 323 (tyrosine 317 in the mouse sequence).
Purification Method Antigen affinity chromatography
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6850
Target Species Human, Mouse
Content And Storage Store at -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.