Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-Tau (Thr181) Mouse anti-Mouse, Human, Clone: 1E7, MilliporeSigma™

Catalog No. MABN388MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN388MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN388MI Supplier MilliporeSigma Supplier No. MABN388
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-phospho Tau (Thr181) antibody, clone 1E7 is an antibody against phospho Tau (Thr181) for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho Tau (Thr181)
Applications Western Blot
Classification Monoclonal
Clone 1.00E+07
Conjugate Unconjugated
Formulation Purified mouse monoclonal in buffer containing PBS without preservatives.
Gene Accession No. NP_001116538
Gene Symbols MAPT, MAPTL, MTBT1, TAU
Host Species Mouse
Immunogen Linear peptide corresponding to human Tau phosphorylated at Thr181.
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.