Learn More
MilliporeSigma™ anti-phospho TDP-43 (Ser409/Ser410), Clone: 1D3, ™
Supplier: MilliporeSigma™ MABN14/DEL

Description
TAR DNA-binding protein 43 (TDP-43) belongs to the hnRNP protein family and plays an important role in transcription, pre-mRNA splicing, mRNA stability and mRNA transport. It is involved in splicing of the apolipoprotein A-II and cystic fibrosis transmembrane gene. This protein is highly expressed in the pancreas, placenta, lung, genital tract and spleen. Mutations in TDP-43 have been associated with amyotrophic lateral sclerosis, frontotemporal dementia, Parkinson's disease and Alzheimer's disease.
Host Species: Rat
Clone: 1D3
Isotype: IgG2a, κ
Cross Reactivity: Human
Immunogen: Ovalbumin-conjugated linear peptide corresponding to human TDP-43 phosphorylated at Ser409/Ser410.
ELISA, Immunohistochemistry, Western Blotting

Specifications
phospho TDP-43 (Ser409/Ser410) | |
1D3 | |
Rat | |
100 μg | |
Human | |
IgG2a κ |
Monoclonal | |
Unconjugated | |
Ovalbumin-conjugated linear peptide corresponding to human TDP-43 phosphorylated at Ser409/Ser410. | |
Primary | |
Purified |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.