Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-Threonine Mouse anti-Human, Mouse, All, Clone: 20H6.1, MilliporeSigma™

Catalog No. 051923MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-192-3MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-192-3MI Supplier MilliporeSigma Supplier No. 051923
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This phospho-Threonine antibody is validated for use in western blotting and immunocytochemistry for the detection of the phospho-Threonine protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-Threonine
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 20H6.1
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2bκ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Symbols THR
Host Species Mouse
Immunogen KLH-conjugated linear peptide corresponding to Threonine.
Purification Method Protein G
Quantity 100 μg
Primary or Secondary Primary
Target Species All species, Human, Mouse
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.