Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-TIE2/TEK Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. ABF131MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABF131MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABF131MI Supplier MilliporeSigma Supplier No. ABF131
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This Anti-phospho TIE2/TEK Antibody (Tyr992) is validated for use in western blotting, immunoprecipitation and peptide inhibition assay for the detection of phospho TIE2/TEK (Tyr992).
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho TIE2/TEK (Tyr992)
Applications Immunoprecipitation, Inhibition Assays, Western Blot
Classification Polyclonal
Concentration 0.25mg/mL
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Symbols TEK; TIE2; VMCM; VMCM1
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to the Protein Kinase Domain of human TIE2/TEK phosphorylated at Tyr992.
Purification Method Affinity Purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_000450
Target Species Human
Content And Storage 2-8°C for 12 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.