Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-TIN2 (Ser295) Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. ABE1314MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABE1314MI 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE1314MI Supplier MilliporeSigma Supplier No. ABE1314
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-phospho TIN2 (Ser295) antibody is an antibody against phospho TIN2 (Ser295) for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho TIN2 (Ser295)
Applications Western Blot
Classification Polyclonal
Concentration 0.25mg/mL
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Symbols TINF2; TIN2
Host Species Rabbit
Immunogen Linear peptide corresponding to human TIN2 .
Purification Method Affinity Purified
Quantity 200 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_001092744
Target Species Human
Content And Storage 2-8°C for 12 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.