Learn More
phospho-TSC2 S1798, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of TSC2.
Supplier: Abnova Corporation PAB8151
Description
Mutations in this gene lead to tuberous sclerosis complex. Its gene product is believed to be a tumor suppressor and is able to stimulate specific GTPases. The protein associates with hamartin in a cytosolic complex, possibly acting as a chaperone for hamartin. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq
Specifications
phospho-TSC2 S1798 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic phosphopeptide of TSC2. | |
In PBS (0.09% sodium azide) | |
FLJ43106/LAM/TSC4 | |
Rabbit | |
400 μL | |
Primary | |
Human | |
Liquid |
Dot Blot, ELISA | |
Unconjugated | |
ELISA (1:1000) Dot Blot (1:500) The optimal working dilution should be determined by the end user. | |
TSC2 | |
TSC2 | |
Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding S1798 of human TSC2. | |
RUO | |
7249 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.