Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-ULK1 (Ser 467) Mouse anti-Human, Clone: 15D12.3, MilliporeSigma™

Catalog No. MABC307MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC307MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABC307MI Supplier MilliporeSigma Supplier No. MABC307
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-phospho-ULK1 (Ser 467) antibody, clone 15D12.3 is validated for use in western blotting for the detection of phospho-ULK1 (Ser 467).
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-ULK1 (Ser 467)
Applications Western Blot
Classification Monoclonal
Clone 15D12.3
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgMλ supernatant in buffer containing PBS with 0.05% sodium azide.
Gene Accession No. NP_003556
Gene Symbols ULK1, KIAA0722
Host Species Mouse
Immunogen KLH-conjugated linear peptide corresponding to human phospho-ULK1 (Ser 467).
Purification Method Protein G purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgGM λ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.