Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Phospho-VASP (Ser239) Mouse anti-Human, Mouse, Rat, Clone: 16C2

Catalog No. 05611MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-611-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-611-MI Supplier MilliporeSigma™ Supplier No. 05611
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-phospho-VASP (Ser239) Antibody, clone 16C2 is validated for use in flow cytometry, western blot, IC for the detection of phospho-VASP (Ser239).
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-VASP (Ser239)
Applications Flow Cytometry, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 16C2
Conjugate Unconjugated
Formulation 100μg of mouse IgG1 from serum-free cell culture supernatant, purified by thiophilic adsorption and size exclusion chromatography. Lyophilized from 1ml of PBS, 0.1% sodium azide, PEG and sucrose. Lyophilized powder. Reconstitute with 100μL H2O (1
Gene Accession No. NM_003370
Gene Symbols VASP
Host Species Mouse
Immunogen phospho-peptide corresponding to amino acids 236-242 of human phospho-VASP (RKV[pS]KQE)
Purification Method Thiophilic adsorption chromatography
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage −20°C for 24 months.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.